Assay Details
Gene Family ID: | MIPF0000022, mir-7 |
Mature miRNA Sequence: | ACAACAAAUCACAGUCUGCCAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-7a-1-3p | MIMAT0000607 |
Stem-loop Details
Rat
More Information
Related Products
miRBase Alias:
Rat: rno-miR-7a-1* (v18)
Rat: rno-miR-7a* (v15)
Rat: rno-miR-7a* (v15)

Genomic Map