Assay Details
Amplicon Length: | 95 |
Chromosome Location: | Chr. 17 - 6675265 - 6675265 [-] on 6675265 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-7a-5p | MIMAT0000606 |
| Rat | rno-miR-7a-1-3p | MIMAT0000607 |
Stem-loop Details
Rat

Genomic Map