Assay Details
Gene Family ID: | MIPF0000178, mir-208 |
Mature miRNA Sequence: | GAGCUUUUGGCCCGGGUUAUAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-208a-5p | MIMAT0026474 |
| Mouse | mmu-miR-208a-5p | MIMAT0017014 |
| Rat | rno-miR-208a-5p | MIMAT0017155 |
| Dasypus novemcinctus | dno-miR-208a-5p | MIMAT0047750 |
| Guinea pig | cpo-miR-208a-5p | MIMAT0047105 |
| Pteropus alecto | pal-miR-208a-5p | MIMAT0040025 |
| View More Rows | ||
| Rhesus monkey | mml-miR-208a-5p | MIMAT0026844 |
Stem-loop Details
Human
Mouse
Rat
Dasypus novemcinctus
Guinea pig
Pteropus alecto
Rhesus monkey

Genomic Map