Assay Details
Amplicon Length: | 95 |
Chromosome Location: | Chr. 6 - 133892902 - 133892902 [-] on 133892902 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-541-5p | MIMAT0003177 |
| Rat | rno-miR-541-3p | MIMAT0017213 |
Stem-loop Details
Rat

Genomic Map