Assay Details
Amplicon Length: | 91 |
Chromosome Location: | Chr. X - 151260542 - 151260542 [+] on 151260542 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-742-3p | MIMAT0005333 |
| Rat | rno-miR-742-5p | MIMAT0017314 |
| Rat | rno-miR-883-3p | MIMAT0005290 |
| Rat | rno-miR-883-5p | MIMAT0017294 |
Stem-loop Details
Rat

Genomic Map