Assay Details
Gene Family ID: | MIPF0000110, mir-329 |
Mature miRNA Sequence: | AAACAUUCGCGGUGCACUUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-543-3p | MIMAT0003175 |
| Goat | chi-miR-543-3p | MIMAT0036275 |
| Gorilla gorilla | ggo-miR-543 | MIMAT0024147 |
Stem-loop Details
Rat
Goat
Gorilla gorilla

Genomic Map