Assay Details
Amplicon Length: | 59 |
Chromosome Location: | Chr. 6 - 133866913 - 133866913 [+] on 133866913 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-543-5p | MIMAT0004787 |
| Rat | rno-miR-543-3p | MIMAT0003175 |
| Rat | rno-miR-666-5p | MIMAT0012855 |
| Rat | rno-miR-666-3p | MIMAT0017371 |
Stem-loop Details
Rat

Genomic Map