Assay Details
Mature miRNA Sequence: | UGAGGUAGUAGUUUGUGCUGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Alligator mississippiensis | ami-let-7i-5p | MIMAT0038016 | |
| Chicken | gga-let-7i | MIMAT0001098 | |
| Cyprinus carpio | ccr-let-7i | MIMAT0026192 | |
| Gorilla gorilla | ggo-let-7i | MIMAT0024083 | |
| Monodelphis domestica | mdo-let-7i-5p | MIMAT0004143 | |
| Sarcophilus harrisii | sha-let-7i | MIMAT0022822 | |
| View More Rows | |||
| Xenopus laevis | xla-let-7i-5p | MIMAT0046407 | |
| Xenopus tropicalis | xtr-let-7i | MIMAT0003647 | |
Stem-loop Details
Alligator mississippiensis
Chicken
Cyprinus carpio
Gorilla gorilla
Monodelphis domestica
Sarcophilus harrisii
Xenopus laevis
Xenopus tropicalis
More Information
Related Products
miRBase Alias:
Human: hsa-let-7i (v9.2)
Mouse: mmu-let-7i (v9.2)
Rat: rno-let-7i (v9.2)
Monodelphis domestica: mdo-let-7i (v19)
Mouse: mmu-let-7i (v9.2)
Rat: rno-let-7i (v9.2)
Monodelphis domestica: mdo-let-7i (v19)

Genomic Map