Assay Details
Mature miRNA Sequence: | UGAGGUAGUAGUUUGUGCUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Alligator mississippiensis | ami-let-7i-5p | MIMAT0038016 |
| Chicken | gga-let-7i | MIMAT0001098 |
| Cyprinus carpio | ccr-let-7i | MIMAT0026192 |
| Gorilla gorilla | ggo-let-7i | MIMAT0024083 |
| Monodelphis domestica | mdo-let-7i-5p | MIMAT0004143 |
| Sarcophilus harrisii | sha-let-7i | MIMAT0022822 |
| View More Rows | ||
| Xenopus laevis | xla-let-7i-5p | MIMAT0046407 |
| Xenopus tropicalis | xtr-let-7i | MIMAT0003647 |
Stem-loop Details
Alligator mississippiensis
Chicken
Cyprinus carpio
Gorilla gorilla
Monodelphis domestica
Sarcophilus harrisii
Xenopus laevis
Xenopus tropicalis
More Information
Related Products
miRBase Alias:
Monodelphis domestica: mdo-let-7i (v19)

Genomic Map