Assay Details
Mature miRNA Sequence: | UGGCAGUGUCUUAGCUGGUUGUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Alligator mississippiensis | ami-miR-34a-5p | MIMAT0038095 | |
| Callithrix jacchus | cja-miR-34a | MIMAT0039496 | |
| Chicken | gga-miR-34a-5p | MIMAT0001173 | |
| Monodelphis domestica | mdo-miR-34a-5p | MIMAT0004096 | |
| Ophiophagus hannah | oha-miR-34a-5p | MIMAT0036907 | |
| Tupaia chinensis | tch-miR-34a-5p | MIMAT0036510 | |
| View More Rows | |||
| Xenopus tropicalis | xtr-miR-34a | MIMAT0003578 | |
Stem-loop Details
Alligator mississippiensis
Callithrix jacchus
Chicken
Monodelphis domestica
Ophiophagus hannah
Tupaia chinensis
Xenopus tropicalis
More Information
Related Products
miRBase Alias:
Human: hsa-miR-34a (v9.2)
Mouse: mmu-miR-34a (v9.2)
Rat: rno-miR-34a (v9.2)
Gallus gallus: gga-miR-34a (v19)
Monodelphis domestica: mdo-miR-34a (v19)
Mouse: mmu-miR-34a (v9.2)
Rat: rno-miR-34a (v9.2)
Gallus gallus: gga-miR-34a (v19)
Monodelphis domestica: mdo-miR-34a (v19)

Genomic Map