Assay Details
Mature miRNA Sequence: | UGGCAGUGUCUUAGCUGGUUGUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Alligator mississippiensis | ami-miR-34a-5p | MIMAT0038095 |
| Callithrix jacchus | cja-miR-34a | MIMAT0039496 |
| Chicken | gga-miR-34a-5p | MIMAT0001173 |
| Monodelphis domestica | mdo-miR-34a-5p | MIMAT0004096 |
| Ophiophagus hannah | oha-miR-34a-5p | MIMAT0036907 |
| Tupaia chinensis | tch-miR-34a-5p | MIMAT0036510 |
| View More Rows | ||
| Xenopus tropicalis | xtr-miR-34a | MIMAT0003578 |
Stem-loop Details
Alligator mississippiensis
Callithrix jacchus
Chicken
Monodelphis domestica
Ophiophagus hannah
Tupaia chinensis
Xenopus tropicalis
More Information
Related Products
miRBase Alias:
Gallus gallus: gga-miR-34a (v19)
Monodelphis domestica: mdo-miR-34a (v19)
Monodelphis domestica: mdo-miR-34a (v19)

Genomic Map