Assay Details
Gene Family ID: | MIPF0000058, mir-205 |
Mature miRNA Sequence: | UCCUUCAUUCCACCGGAGUCUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-205-5p | MIMAT0000266 | |
| Mouse | mmu-miR-205-5p | MIMAT0000238 | |
| Alligator mississippiensis | ami-miR-205a-5p | MIMAT0038228 | |
| Anolis carolinensis | aca-miR-205a | MIMAT0021855 | |
| Astatotilapia burtoni | abu-miR-205 | MIMAT0042062 | |
| Ateles geoffroyi | age-miR-205 | MIMAT0002542 | |
| View More Rows | |||
| Bovine | bta-miR-205 | MIMAT0003545 | |
| Chicken | gga-miR-205a | MIMAT0001184 | |
| Chrysemys picta | cpi-miR-205a-5p | MIMAT0037830 | |
| Columba livia | cli-miR-205a-5p | MIMAT0038584 | |
| Cyprinus carpio | ccr-miR-205 | MIMAT0026263 | |
| Dog | cfa-miR-205 | MIMAT0009845 | |
| Fugu rubripes | fru-miR-205 | MIMAT0002965 | |
| Gorilla gorilla | ggo-miR-205 | MIMAT0002541 | |
| Horse | eca-miR-205 | MIMAT0012962 | |
| Lagothrix lagotricha | lla-miR-205 | MIMAT0002545 | |
| Macaca nemestrina | mne-miR-205 | MIMAT0002546 | |
| Metriaclima zebra | mze-miR-205 | MIMAT0042296 | |
| Monodelphis domestica | mdo-miR-205a | MIMAT0012749 | |
| Neolamprologus brichardi | nbr-miR-205 | MIMAT0042484 | |
| Oreochromis niloticus | oni-miR-205 | MIMAT0042668 | |
| Pan paniscus | ppa-miR-205 | MIMAT0002543 | |
| Pan troglodytes | ptr-miR-205 | MIMAT0002544 | |
| Pig | ssc-miR-205 | MIMAT0002146 | |
| Pongo pygmaeus | ppy-miR-205 | MIMAT0015798 | |
| Pundamilia nyererei | pny-miR-205 | MIMAT0042873 | |
| Python bivittatus | pbv-miR-205a-5p | MIMAT0039009 | |
| Rabbit | ocu-miR-205-5p | MIMAT0048325 | |
| Rhesus monkey | mml-miR-205 | MIMAT0006236 | |
| Salmo salar | ssa-miR-205b-5p | MIMAT0032495 | |
| Tetraodon nigroviridis | tni-miR-205 | MIMAT0002966 | |
| Xenopus laevis | xla-miR-205 | MIMAT0011142 | |
| Xenopus tropicalis | xtr-miR-205a | MIMAT0003697 | |
| Zebrafish | dre-miR-205-5p | MIMAT0001280 | |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-205 |
| Stem-loop Accession # | MI0000285 |
| Stem-loop Sequence | AAAGAUCCUCAGACAAUCCAUGUGCUUCUCUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUACCCAACCAGAUUUCAGUGGAGUGAAGUUCAGGAGGCAUGGAGCUGACA |
| Chromosome Location | Chr. 1 - 209432133 - 209432242 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000266
miRBase ID: hsa-miR-205-5p Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: Chr. 1 - 209432133 - 209432242 [+] on Build GRCh38 |
| Stem-loop ID | mmu-mir-205 |
| Stem-loop Accession # | MI0000248 |
| Stem-loop Sequence | CUCUUGUCCUUCAUUCCACCGGAGUCUGUCUUAUGCCAACCAGAUUUCAGUGGAGUGAAGCUCAGGAG |
| Chromosome Location | Chr. 1 - 193507463 - 193507530 [-] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000238
miRBase ID: mmu-miR-205-5p Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: Chr. 1 - 193507463 - 193507530 [-] on Build GRCm38 |
| Stem-loop ID | ami-mir-205a |
| Stem-loop Accession # | MI0029791 |
| Stem-loop Sequence | UCCAUGGAUUCUGUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUACCUAAUCAGAUUUCAGUGGAGUGAAGCACAAGAGACAUGGAGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038228
miRBase ID: ami-miR-205a-5p Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | aca-mir-205a |
| Stem-loop Accession # | MI0018808 |
| Stem-loop Sequence | GUUCUAUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUAUCUAAUCAGAUUUCAGUGGAGUGAAGCACAAGAGACA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0021855
miRBase ID: aca-miR-205a Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | abu-mir-205-1 |
| Stem-loop Accession # | MI0033757 |
| Stem-loop Sequence | UCCUUCAUUCCACCGGAGUCUGUGUAAAGUGCUAAUCAGAUUUCAGUGGUUUGAAGAGUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042062
miRBase ID: abu-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | abu-mir-205-2 |
| Stem-loop Accession # | MI0033899 |
| Stem-loop Sequence | UCCUUCAUUCCACCGGAGUCUGUAUCUGUAUUUGACCAGAUUUCAGUGGUGUGAAGUGUAAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042062
miRBase ID: abu-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | age-mir-205 |
| Stem-loop Accession # | MI0002845 |
| Stem-loop Sequence | AAAGAUCCUCGGGCAAUCCAUGUGCUUCUCUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUACCCAACCAGAUUUCAGUGGAGUGAAGUCAGGAGGCAUGGAGCUGACG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002542
miRBase ID: age-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | bta-mir-205 |
| Stem-loop Accession # | MI0004759 |
| Stem-loop Sequence | CUCUUGUCCUUCAUUCCACCGGAGUCUGUCUCGUACCCAACCAGAUUUCAGUGGAGUGAAGUUCAGGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003545
miRBase ID: bta-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | gga-mir-205a |
| Stem-loop Accession # | MI0001267 |
| Stem-loop Sequence | GACAAUCCAUGGGUUCUGUUGUCCUUCAUUCCACCGGAGUCUGUCUCGUACCUAACCAGAUUUCAGUGGAGUGAAGCACAAGAGACAUGGAGAUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001184
miRBase ID: gga-miR-205a Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | cpi-mir-205a |
| Stem-loop Accession # | MI0029531 |
| Stem-loop Sequence | AUCCAUGAAUUCUGUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUAACUAAUCAGAUUUCAGUGGAGUGAAGCACAAGAGACAUGGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0037830
miRBase ID: cpi-miR-205a-5p Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | cli-mir-205a |
| Stem-loop Accession # | MI0030011 |
| Stem-loop Sequence | CAAUCCAUGGGUUCUGUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUACCUAAACCAGAUUUCAGUGGAGUGAAGUACGAGAGACAUGGAGAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038584
miRBase ID: cli-miR-205a-5p Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | ccr-mir-205 |
| Stem-loop Accession # | MI0023365 |
| Stem-loop Sequence | CUGUGAAUUCUAUCCUUCAUUCCACCGGAGUCUGUGUAGCUGUCCAAUCAGAUUUCAGUGGUGUGAAGUGUAGGAGACAUGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0026263
miRBase ID: ccr-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | cfa-mir-205 |
| Stem-loop Accession # | MI0010340 |
| Stem-loop Sequence | CUCUUGUCCUUCAUUCCACCGGAGUCUGUCUCGUACCCAACCAGAUUUCAGUGGAGUGAAGUUCAGGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0009845
miRBase ID: cfa-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | fru-mir-205 |
| Stem-loop Accession # | MI0003281 |
| Stem-loop Sequence | UGUGUUCUAUCCUUCAUUCCACCGGAGUCUGUAUGUGUAUUUAACCAGAUUUCAGUGGUGUGAAGUGUAAGAGACAUGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002965
miRBase ID: fru-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | ggo-mir-205 |
| Stem-loop Accession # | MI0002844 |
| Stem-loop Sequence | AAAGAUCCUCAGACAAUCCAUGUGCUUCUCUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUACCCAACCAGAUUUCAGUGGAGUGAAGUUCAGGAGGCAUGGAGCUGACA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002541
miRBase ID: ggo-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | eca-mir-205 |
| Stem-loop Accession # | MI0012713 |
| Stem-loop Sequence | CUCUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUACCCAACCAGAUUUCAGUGGAGUGAAGUUCAGGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0012962
miRBase ID: eca-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | lla-mir-205 |
| Stem-loop Accession # | MI0002848 |
| Stem-loop Sequence | AAAGAUCCUCGGGCAAUCCAUGUGCUUCUCUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUACCCAACCAGAUUUCAGUGGAGUGAAGUUCAGGAGGCAUGGAGCUGACG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002545
miRBase ID: lla-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | mne-mir-205 |
| Stem-loop Accession # | MI0002849 |
| Stem-loop Sequence | AAAGAUCCUCAGGCAAUCCAUGUGCUUCUCUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUACCCAACCAGAUUUCAGUGGAGUGAAGUUCAGGAGGCAUGGAGCUGACG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002546
miRBase ID: mne-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | mze-mir-205-1 |
| Stem-loop Accession # | MI0034053 |
| Stem-loop Sequence | UCCUUCAUUCCACCGGAGUCUGUGUAAAGUGCUAAUCAGAUUUCAGUGGUUUGAAGAGUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042296
miRBase ID: mze-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | mze-mir-205-2 |
| Stem-loop Accession # | MI0034191 |
| Stem-loop Sequence | UCCUUCAUUCCACCGGAGUCUGUAUCUGUAUUUGACCAGAUUUCAGUGGUGUGAAGUGUAAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042296
miRBase ID: mze-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | mdo-mir-205a |
| Stem-loop Accession # | MI0012506 |
| Stem-loop Sequence | CUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUAUCUAAUCAGAUUUCAGUGGAGUGAAGCAUAAGAGGCAUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0012749
miRBase ID: mdo-miR-205a Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | nbr-mir-205-1 |
| Stem-loop Accession # | MI0034313 |
| Stem-loop Sequence | UCCUUCAUUCCACCGGAGUCUGUGUAAAGUGCUAAUCAGAUUUCAGUGGUUUGAAGAGUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042484
miRBase ID: nbr-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | nbr-mir-205-2 |
| Stem-loop Accession # | MI0034447 |
| Stem-loop Sequence | UCCUUCAUUCCACCGGAGUCUGUAUCUGUAUUUGACCAGAUUUCAGUGGUGUGAAGUGUAAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042484
miRBase ID: nbr-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | oni-mir-205-1 |
| Stem-loop Accession # | MI0034567 |
| Stem-loop Sequence | UCCUUCAUUCCACCGGAGUCUGUGUAAAGUGCCAAUCAGAUUUCAGUGGUUUGAAGAGUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042668
miRBase ID: oni-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | oni-mir-205-2 |
| Stem-loop Accession # | MI0034703 |
| Stem-loop Sequence | UCCUUCAUUCCACCGGAGUCUGUAUCUGUAUUUGACCAGAUUUCAGUGGUGUGAAGUGUAAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042668
miRBase ID: oni-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | ppa-mir-205 |
| Stem-loop Accession # | MI0002846 |
| Stem-loop Sequence | AAAGAUCCUCAGACAAUCCAUGUGCUUCUCUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUACCCAACCAGAUUUCAGUGGAGUGAAGUUCAGGAGGCAUGGAGCUGACA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002543
miRBase ID: ppa-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | ptr-mir-205 |
| Stem-loop Accession # | MI0002847 |
| Stem-loop Sequence | AAAGAUCCUCAGACAAUCCAUGUGCUUCUCUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUACCCAACCAGAUUUCAGUGGAGUGAAGUUCAGGAGGCAUGGAGCUGACA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002544
miRBase ID: ptr-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | ssc-mir-205 |
| Stem-loop Accession # | MI0002440 |
| Stem-loop Sequence | CUCUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUACCCAACCAGAUUUCAGUGGAGUGAAGCUCAGGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002146
miRBase ID: ssc-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | ppy-mir-205 |
| Stem-loop Accession # | MI0014863 |
| Stem-loop Sequence | AAAGAUCCUCAGACAAUCCAUGUGCUUCUCUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUACCCAACCAGAUUUCAGUGGAGUGAAGUUCAGGAGGCAUGGAGCUGAC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015798
miRBase ID: ppy-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | pny-mir-205-1 |
| Stem-loop Accession # | MI0034829 |
| Stem-loop Sequence | UCCUUCAUUCCACCGGAGUCUGUGUAAAGUGCUAAUCAGAUUUCAGUGGUUUGAAGAGUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042873
miRBase ID: pny-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | pny-mir-205-2 |
| Stem-loop Accession # | MI0034956 |
| Stem-loop Sequence | UCCUUCAUUCCACCGGAGUCUGUAUCUGUAUUUGACCAGAUUUCAGUGGUGUGAAGUGUAAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042873
miRBase ID: pny-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | pbv-mir-205a |
| Stem-loop Accession # | MI0030281 |
| Stem-loop Sequence | CCAUGCAUUCUGUUGUCCUUCAUUCCACCGGAGUCUGUCGAAUACCUAAUCAGAUUUCAGUGGCGUGAAGUACAUGAGACAUGGAGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039009
miRBase ID: pbv-miR-205a-5p Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | ocu-mir-205 |
| Stem-loop Accession # | MI0039342 |
| Stem-loop Sequence | UCCUUCAUUCCACCGGAGUCUGUCUCAUACCCAACCAGAUUUCAGUGGAGUGAAGUUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048325
miRBase ID: ocu-miR-205-5p Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | mml-mir-205 |
| Stem-loop Accession # | MI0007666 |
| Stem-loop Sequence | AAAGAUCCUCAGGCAAUCCAUGUGCUUCUCUUGUCCUUCAUUCCACCGGAGUCUGUCUCAUACCCAACCAGAUUUCAGUGGAGUGAAGUUCAGGAGGCAUGGAGCUGACG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006236
miRBase ID: mml-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | ssa-mir-205b-1 |
| Stem-loop Accession # | MI0026596 |
| Stem-loop Sequence | UCCUUCAUUCCACCGGAGUCUGUGUCAUGAACCAAUCAGAUUUCAGUGGUAUGAAGAGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032495
miRBase ID: ssa-miR-205b-5p Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | ssa-mir-205b-2 |
| Stem-loop Accession # | MI0026597 |
| Stem-loop Sequence | UCCUUCAUUCCACCGGAGUCUGUGUCAUGAACCAUUCAGAUUUCAGUGGUGUGAAGAGUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032495
miRBase ID: ssa-miR-205b-5p Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | ssa-mir-205b-3 |
| Stem-loop Accession # | MI0026598 |
| Stem-loop Sequence | UCCUUCAUUCCACCGGAGUCUGUGCCUUUAUCCAAACAGAUUUCAGUGAAGUGAAGUGUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032495
miRBase ID: ssa-miR-205b-5p Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | ssa-mir-205b-4 |
| Stem-loop Accession # | MI0026599 |
| Stem-loop Sequence | UCCUUCAUUCCACCGGAGUCUGUGUCUUUAUUCAACCAGAUUUCUGUGAAAUGAAGUGUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032495
miRBase ID: ssa-miR-205b-5p Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | tni-mir-205 |
| Stem-loop Accession # | MI0003282 |
| Stem-loop Sequence | UAUCCUUCAUUCCACCGGAGUCUGUAACUGUUUUUAACCAGAUUUCAGUGGUGUGAAGUGUAAGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002966
miRBase ID: tni-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | xla-mir-205 |
| Stem-loop Accession # | MI0008380 |
| Stem-loop Sequence | CCACGUGUCCUGCUGUCCUUCAUUCCACCGGAGUCUGUCUCAUACAUAAUCAGAUUUCAGUAGAGUGAAGCACAAGAGGCAUGUAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0011142
miRBase ID: xla-miR-205 Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | xtr-mir-205a |
| Stem-loop Accession # | MI0004950 |
| Stem-loop Sequence | CCACGUGUCCUGCUGUCCUUCAUUCCACCGGAGUCUGUCUCAUACAUAAUCAGAUUUCAGUGGAGUGAAGCACAAGAGGCAUGUAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003697
miRBase ID: xtr-miR-205a Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
| Stem-loop ID | dre-mir-205 |
| Stem-loop Accession # | MI0001378 |
| Stem-loop Sequence | AAACUACUGUGCAUUCUAUCCUUCAUUCCACCGGAGUCUGUGUAGUUGUUCAAUCAGAUUUCAGUGGUGUGAAGUGUAGGAAACACGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001280
miRBase ID: dre-miR-205-5p Mature miRNA Sequence: UCCUUCAUUCCACCGGAGUCUG Chromosome Location: NA |
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products
miRBase Alias:
Human: hsa-miR-205 (v17)
Mouse: mmu-miR-205 (v17)
Rat: rno-miR-205 (v15)
Danio rerio: dre-miR-205 (v20)
Mouse: mmu-miR-205 (v17)
Rat: rno-miR-205 (v15)
Danio rerio: dre-miR-205 (v20)

Genomic Map