Assay Details
Amplicon Length: | 77 |
Chromosome Location: | Chr. 1 - 193507058 - 193507058 [-] on 193507058 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-205-3p | MIMAT0017003 |
| Mouse | mmu-miR-205-5p | MIMAT0000238 |
Stem-loop Details
Mouse

Genomic Map