Assay Details
Gene Family ID: | MIPF0000163, mir-320 |
Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGGGCGAA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pig | ssc-miR-320 | MIMAT0013878 |
Stem-loop Details
Pig
More Information
Related Products
miRBase Alias:
Human: hsa-miR-320 (v9.2)
Mouse: mmu-miR-320 (v9.2)
Rat: rno-miR-320 (v9.2)
Mouse: mmu-miR-320 (v9.2)
Rat: rno-miR-320 (v9.2)

Genomic Map