Assay Details
Gene Family ID: | MIPF0000163, mir-320 |
Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGGGCGAA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pig | ssc-miR-320 | MIMAT0013878 |
Stem-loop Details
Pig
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | |
|---|---|
| miRBase Accession Number: | MI0013097 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGGGCGAA |
| Species: | Pig |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000163, mir-320 |
Mature miRNA Sequence: | AAAAGCUGGGUUGAGAGGGCGAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pig | ssc-miR-320 | MIMAT0013878 |