Assay Details
Gene Family ID: | MIPF0000500, mir-373 |
Mature miRNA Sequence: | ACUCAAAAUGGGGGCGCUUUCC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-373-5p | MIMAT0000725 |
| Pan troglodytes | ptr-miR-373 | MIMAT0012802 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map