Assay Details
Amplicon Length: | 81 |
Chromosome Location: | Chr. 19 - 53789206 - 53789206 [+] on 53789206 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-373-3p | MIMAT0000726 |
| Human | hsa-miR-373-5p | MIMAT0000725 |
Stem-loop Details
Human

Genomic Map