Assay Details
Gene Family ID: | MIPF0000288, mir-374 |
Mature miRNA Sequence: | UUAUAAUACAACCUGAUAAGUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-374a-5p | MIMAT0000727 | |
| Bovine | bta-miR-374a | MIMAT0004342 | |
| Dasypus novemcinctus | dno-miR-374a-5p | MIMAT0047846 | |
| Guinea pig | cpo-miR-374-5p | MIMAT0047205 | |
| Horse | eca-miR-374a | MIMAT0013213 | |
| Pan troglodytes | ptr-miR-374a | MIMAT0008123 | |
| View More Rows | |||
| Pig | ssc-miR-374a-5p | MIMAT0013913 | |
| Pongo pygmaeus | ppy-miR-374a | MIMAT0015862 | |
| Rabbit | ocu-miR-374a-5p | MIMAT0048406 | |
| Rhesus monkey | mml-miR-374a-5p | MIMAT0006300 | |
| Sheep | oar-miR-374a | MIMAT0030062 | |
| Tupaia chinensis | tch-miR-374a-5p | MIMAT0036589 | |
Stem-loop Details
Human
Bovine
Dasypus novemcinctus
Guinea pig
Horse
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey
Sheep
Tupaia chinensis
More Information
Related Products
miRBase Alias:
Human: hsa-miR-374 (v9.2)
Human: hsa-miR-374a (v17)
Bos taurus: bta-miR-374 (v11)
Macaca mulatta: mml-miR-374a (v19)
Staphylococcus schleiferi: ssc-miR-374a (v18)
Human: hsa-miR-374a (v17)
Bos taurus: bta-miR-374 (v11)
Macaca mulatta: mml-miR-374a (v19)
Staphylococcus schleiferi: ssc-miR-374a (v18)

Genomic Map