Assay Details
Amplicon Length: | 74 |
Chromosome Location: | Chr. X - 74287501 - 74287501 [-] on 74287501 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-374a-3p | MIMAT0004688 |
| Human | hsa-miR-374a-5p | MIMAT0000727 |
Stem-loop Details
Human

Genomic Map