Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UAUGUAAUAUGGUCCACAUCUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-380-3p | MIMAT0000735 |
| Pan troglodytes | ptr-miR-380 | MIMAT0008132 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map