Assay Details
Amplicon Length: | 71 |
Chromosome Location: | Chr. 14 - 101025417 - 101025417 [+] on 101025417 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1197 | MIMAT0005955 |
| Human | hsa-miR-323a-5p | MIMAT0004696 |
| Human | hsa-miR-323a-3p | MIMAT0000755 |
| Human | hsa-miR-380-5p | MIMAT0000734 |
| Human | hsa-miR-380-3p | MIMAT0000735 |
Stem-loop Details
Human
More Information
Related Products
miRBase Alias:
Human: hsa-mir-380 (v21)
Human: hsa-mir-323a (v21)
Human: hsa-mir-323a (v21)

Genomic Map