Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UGGUUGACCAUAGAACAUGCGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-380-5p | MIMAT0000734 |
| Bovine | bta-miR-380-5p | MIMAT0003803 |
Stem-loop Details
Human
Bovine
More Information
Related Products
miRBase Alias:
Human: hsa-miR-380* (v17)
Mouse: mmu-miR-380-5p (v9.2)
Mouse: mmu-miR-380-5p (v9.2)

Genomic Map