Assay Details
Gene Family ID: | MIPF0000186, mir-299 |
Mature miRNA Sequence: | UAUGUGGGAUGGUAAACCGCUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-299-3p | MIMAT0000687 |
| Dasypus novemcinctus | dno-miR-299-3p | MIMAT0047786 |
| Horse | eca-miR-299 | MIMAT0013130 |
Stem-loop Details
Human
Dasypus novemcinctus
Horse

Genomic Map