Assay Details
Amplicon Length: | 62 |
Chromosome Location: | Chr. 14 - 101023995 - 101023995 [-] on 101023995 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-299-3p | MIMAT0000687 |
| Human | hsa-miR-299-5p | MIMAT0002890 |
Stem-loop Details
Human

Genomic Map