Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGAUCGACCGUGUUAUAUUCGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-369-5p | MIMAT0001621 |
| Mouse | mmu-miR-369-5p | MIMAT0003185 |
| Rat | rno-miR-369-5p | MIMAT0003206 |
| Cricetulus griseus | cgr-miR-369-5p | MIMAT0023937 |
| Pongo pygmaeus | ppy-miR-369-5p | MIMAT0015855 |
| Rhesus monkey | mml-miR-369-5p | MIMAT0006293 |
Stem-loop Details
Human
Mouse
Rat
Cricetulus griseus
Pongo pygmaeus
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products

Genomic Map