Assay Details
Amplicon Length: | 75 |
Chromosome Location: | Chr. 14 - 101065146 - 101065146 [-] on 101065146 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-369-3p | MIMAT0000721 |
| Human | hsa-miR-369-5p | MIMAT0001621 |
| Human | hsa-miR-409-5p | MIMAT0001638 |
| Human | hsa-miR-409-3p | MIMAT0001639 |
| Human | hsa-miR-412-3p | MIMAT0002170 |
| Human | hsa-miR-412-5p | MIMAT0026557 |
Stem-loop Details
Human
More Information
Related Products
miRBase Alias:
Human: hsa-mir-369 (v21)
Human: hsa-mir-412 (v21)
Human: hsa-mir-412 (v21)

Genomic Map