Assay Details
Gene Family ID: | MIPF0000139, mir-500 |
Mature miRNA Sequence: | AUGCACCUGGGCAAGGAUUCUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-500a-3p | MIMAT0002871 |
Stem-loop Details
Human
More Information
Related Products
miRBase Alias:
Human: hsa-miR-500a* (v17)
Human: hsa-miR-500 (v9.2)
Human: hsa-miR-500* (v15)
Human: hsa-miR-500 (v9.2)
Human: hsa-miR-500* (v15)

Genomic Map