Assay Details
Amplicon Length: | 94 |
Chromosome Location: | Chr. X - 50008131 - 50008131 [-] on 50008131 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-500a-5p | MIMAT0004773 |
| Human | hsa-miR-500a-3p | MIMAT0002871 |
Stem-loop Details
Human

Genomic Map