Assay Details
Gene Family ID: | MIPF0000068, mir-290 |
Mature miRNA Sequence: | AAAGUGCUUCCACUUUGUGUGCC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-291a-3p | MIMAT0000895 |
Stem-loop Details
Rat
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-291a-3p (v9.2)
Mouse: mmu-miR-291-3p (v7.1)
Rat: rno-miR-291-3p (v9.2)
Mouse: mmu-miR-291-3p (v7.1)
Rat: rno-miR-291-3p (v9.2)

Genomic Map