Assay Details
Amplicon Length: | 102 |
Chromosome Location: | Chr. 1 - 64537953 - 64537953 [+] on 64537953 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-291a-5p | MIMAT0000894 |
| Rat | rno-miR-291a-3p | MIMAT0000895 |
| Rat | rno-miR-292-5p | MIMAT0000896 |
| Rat | rno-miR-292-3p | MIMAT0000897 |
Stem-loop Details
Rat

Genomic Map