Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | CUCCAGAGGGAUGCACUUUCU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-525-5p | MIMAT0002838 |
| Gorilla gorilla | ggo-miR-525-5p | MIMAT0036412 |
| Pongo pygmaeus | ppy-miR-525-5p | MIMAT0015982 |
| Rhesus monkey | mml-miR-525 | MIMAT0006402 |
Stem-loop Details
Human
Gorilla gorilla
Pongo pygmaeus
Rhesus monkey

Genomic Map