Assay Details
Amplicon Length: | 76 |
Chromosome Location: | Chr. 19 - 53697139 - 53697139 [-] on 53697139 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-525-5p | MIMAT0002838 |
Human | hsa-miR-525-3p | MIMAT0002839 |
Stem-loop Details
Human
