Assay Details
Gene Family ID: | MIPF0000063, mir-192 |
Mature miRNA Sequence: | AUGACCUAUGAUUUGACAGAC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-215-5p | MIMAT0000904 |
Stem-loop Details
Mouse
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-215 (v17)
Rat: rno-miR-215 (v15)
Rat: rno-miR-215 (v15)
Citations (1)
NAK is recruited to the TNFR1 complex in a TNFalpha-dependent manner and mediates the production of RANTES: identification of endogenous TNFR-interacting proteins by a proteomic approach.
- Authors
- Kuai J, Wooters J, Hall JP, Rao VR, Nickbarg E, Li B, Chatterjee-Kishore M, Qiu Y, Lin LL
- Journal
- J Biol Chem - Page No 53266 (Fri Dec 17 00:00:00 UTC 2004)
- Volume (Year)
- 279 (2004)
- Product Usage
- -

Genomic Map