Assay Details
Amplicon Length: | 63 |
Chromosome Location: | Chr. 1 - 185313983 - 185313983 [-] on 185313983 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-215-3p | MIMAT0017169 |
| Mouse | mmu-miR-215-5p | MIMAT0000904 |
Stem-loop Details
Mouse

Genomic Map