Assay Details
Gene Family ID: | MIPF0000053, mir-22 |
Mature miRNA Sequence: | AGUUCUUCAGUGGCAAGCUUU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Cricetulus griseus | cgr-miR-22-5p | MIMAT0023856 |
Goat | chi-miR-22-5p | MIMAT0036075 |
Xenopus tropicalis | xtr-miR-22-5p | MIMAT0003671 |
Stem-loop Details
Cricetulus griseus
Goat
Xenopus tropicalis

More Information
Related Products
miRBase Alias:
Rat: rno-miR-22* (v9.2)
Xenopus tropicalis: xtr-miR-22* (v18)
Xenopus tropicalis: xtr-miR-22* (v18)
