Assay Details
Gene Family ID: | MIPF0000053, mir-22 |
Mature miRNA Sequence: | AGUUCUUCAGUGGCAAGCUUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Cricetulus griseus | cgr-miR-22-5p | MIMAT0023856 |
| Goat | chi-miR-22-5p | MIMAT0036075 |
| Xenopus tropicalis | xtr-miR-22-5p | MIMAT0003671 |
Stem-loop Details
Cricetulus griseus
Goat
Xenopus tropicalis
More Information
Related Products
miRBase Alias:
Rat: rno-miR-22* (v9.2)
Xenopus tropicalis: xtr-miR-22* (v18)
Xenopus tropicalis: xtr-miR-22* (v18)

Genomic Map