Assay Details
Gene Family ID: | MIPF0000555, mir-567 |
Mature miRNA Sequence: | AGUAUGUUCUUCCAGGACAGAAC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-567 | MIMAT0003231 |
| Pan troglodytes | ptr-miR-567 | MIMAT0008245 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map