Assay Details
Amplicon Length: | 64 |
Chromosome Location: | Chr. 3 - 112112602 - 112112602 [-] on 112112602 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-567 | MIMAT0003231 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 64 |
Chromosome Location: | Chr. 3 - 112112602 - 112112602 [-] on 112112602 Build GRCh38 |
Species: | Human |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-567 | MIMAT0003231 |