Assay Details
Gene Family ID: | MIPF0000360, mir-551 |
Mature miRNA Sequence: | GCGACCCAUACUUGGUUUCAG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-551b-3p | MIMAT0003233 | |
Mouse | mmu-miR-551b-3p | MIMAT0003890 | |
Anolis carolinensis | aca-miR-551 | MIMAT0021982 | |
Callithrix jacchus | cja-miR-551 | MIMAT0039411 | |
Chicken | gga-miR-551-3p | MIMAT0007290 | |
Chrysemys picta | cpi-miR-551-2-3p | MIMAT0037905 | |
View More Rows | |||
Dasypus novemcinctus | dno-miR-551b-3p | MIMAT0047977 | |
Dog | cfa-miR-551b | MIMAT0009913 | |
Gorilla gorilla | ggo-miR-551b | MIMAT0024188 | |
Guinea pig | cpo-miR-551-3p | MIMAT0047348 | |
Horse | eca-miR-551b | MIMAT0013094 | |
Monodelphis domestica | mdo-miR-551b-3p | MIMAT0012764 | |
Ornithorhynchus anatinus | oan-miR-551-3p | MIMAT0007130 | |
Pan troglodytes | ptr-miR-551b | MIMAT0008232 | |
Pongo pygmaeus | ppy-miR-551b | MIMAT0015998 | |
Python bivittatus | pbv-miR-551b-3p | MIMAT0039069 | |
Rabbit | ocu-miR-551b-3p | MIMAT0048530 | |
Rhesus monkey | mml-miR-551b-3p | MIMAT0006420 | |
Taeniopygia guttata | tgu-miR-551-3p | MIMAT0014541 |
Stem-loop Details
Human
Mouse
Anolis carolinensis
Callithrix jacchus
Chicken
Chrysemys picta
Dasypus novemcinctus
Dog
Gorilla gorilla
Guinea pig
Horse
Monodelphis domestica
Ornithorhynchus anatinus
Pan troglodytes
Pongo pygmaeus
Python bivittatus
Rabbit
Rhesus monkey
Taeniopygia guttata

More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-551b (v17)
Mouse: mmu-miR-551b (v17)
Gallus gallus: gga-miR-551 (v18)
Macaca mulatta: mml-miR-551b (v19)
Monodelphis domestica: mdo-miR-551b (v19)
Ochrobactrum anthropi: oan-miR-551 (v18)
Trabulsiella guamensis ATCC 49490: tgu-miR-551 (v18)
Mouse: mmu-miR-551b (v17)
Gallus gallus: gga-miR-551 (v18)
Macaca mulatta: mml-miR-551b (v19)
Monodelphis domestica: mdo-miR-551b (v19)
Ochrobactrum anthropi: oan-miR-551 (v18)
Trabulsiella guamensis ATCC 49490: tgu-miR-551 (v18)
