Assay Details
Amplicon Length: | 90 |
Chromosome Location: | Chr. 3 - 168551552 - 168551552 [-] on 168551552 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-551b-5p | MIMAT0004794 |
| Human | hsa-miR-551b-3p | MIMAT0003233 |
Stem-loop Details
Human

Genomic Map