Assay Details
Gene Family ID: | MIPF0000340, mir-592 |
Mature miRNA Sequence: | UUGUGUCAAUAUGCGAUGAUGU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-592 | MIMAT0003260 | |
Eptesicus fuscus | efu-miR-592 | MIMAT0035159 | |
Goat | chi-miR-592 | MIMAT0036280 | |
Horse | eca-miR-592 | MIMAT0012946 | |
Pan troglodytes | ptr-miR-592 | MIMAT0008263 | |
Pongo pygmaeus | ppy-miR-592 | MIMAT0016035 | |
View More Rows | |||
Rhesus monkey | mml-miR-592-5p | MIMAT0006452 | |
Tupaia chinensis | tch-miR-592 | MIMAT0036478 |
Stem-loop Details
Human
Eptesicus fuscus
Goat
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
Tupaia chinensis
