Assay Details
Gene Family ID: | MIPF0000340, mir-592 |
Mature miRNA Sequence: | UUGUGUCAAUAUGCGAUGAUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-592 | MIMAT0003260 |
| Eptesicus fuscus | efu-miR-592 | MIMAT0035159 |
| Goat | chi-miR-592 | MIMAT0036280 |
| Horse | eca-miR-592 | MIMAT0012946 |
| Pan troglodytes | ptr-miR-592 | MIMAT0008263 |
| Pongo pygmaeus | ppy-miR-592 | MIMAT0016035 |
| View More Rows | ||
| Rhesus monkey | mml-miR-592-5p | MIMAT0006452 |
| Tupaia chinensis | tch-miR-592 | MIMAT0036478 |
Stem-loop Details
Human
Eptesicus fuscus
Goat
Horse
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
Tupaia chinensis

Genomic Map