Assay Details
Gene Family ID: | MIPF0000550, mir-627 |
Mature miRNA Sequence: | GUGAGUCUCUAAGAAAAGAGGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-627-5p | MIMAT0003296 |
| Pan troglodytes | ptr-miR-627 | MIMAT0008286 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map