Assay Details
Amplicon Length: | 89 |
Chromosome Location: | Chr. 15 - 42199973 - 42199973 [-] on 42199973 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-627-5p | MIMAT0003296 |
| Human | hsa-miR-627-3p | MIMAT0026623 |
Stem-loop Details
Human

Genomic Map