Assay Details
Gene Family ID: | MIPF0000288, mir-374 |
Mature miRNA Sequence: | GGUUGUAUUAUCAUUGUCCGAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-374b-3p | MIMAT0003728 |
Stem-loop Details
Mouse
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-374* (v17)
Mouse: mmu-miR-374-3p (v18)
Mouse: mmu-miR-374-3p (v18)

Genomic Map