Assay Details
Amplicon Length: | 82 |
Chromosome Location: | Chr. X - 103573416 - 103573416 [-] on 103573416 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-374b-5p | MIMAT0003727 |
| Mouse | mmu-miR-374b-3p | MIMAT0003728 |
| Mouse | mmu-miR-374c-3p | MIMAT0014954 |
| Mouse | mmu-miR-374c-5p | MIMAT0014953 |
Stem-loop Details
Mouse

Genomic Map