Assay Details
Gene Family ID: | MIPF0000056, mir-148 |
Mature miRNA Sequence: | GAAGUUCUGUUAUACACUCAGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-148b-5p | MIMAT0004645 |
| Horse | eca-miR-148b-5p | MIMAT0012971 |
Stem-loop Details
Rat
Horse

Genomic Map