Assay Details
Amplicon Length: | 54 |
Chromosome Location: | Chr. 7 - 144911790 - 144911790 [-] on 144911790 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-148b-3p | MIMAT0000579 |
| Rat | rno-miR-148b-5p | MIMAT0004645 |
Stem-loop Details
Rat

Genomic Map