Assay Details
Gene Family ID: | MIPF0000209, mir-362 |
Mature miRNA Sequence: | AACACACCUAUUCAAGGAUUCA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-362-3p | MIMAT0004683 | |
| Dasypus novemcinctus | dno-miR-362-3p | MIMAT0047832 | |
| Gorilla gorilla | ggo-miR-362 | MIMAT0024241 | |
| Guinea pig | cpo-miR-362-3p | MIMAT0047191 | |
| Horse | eca-miR-362-3p | MIMAT0013211 | |
| Pan troglodytes | ptr-miR-362 | MIMAT0008116 | |
| View More Rows | |||
| Pongo pygmaeus | ppy-miR-362-3p | MIMAT0015851 | |
| Rabbit | ocu-miR-362-3p | MIMAT0048396 | |
| Rhesus monkey | mml-miR-362-3p | MIMAT0006289 | |
Stem-loop Details
Human
Dasypus novemcinctus
Gorilla gorilla
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey

Genomic Map