Assay Details
Amplicon Length: | 59 |
Chromosome Location: | Chr. X - 50009162 - 50009162 [-] on 50009162 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-362-3p | MIMAT0004683 |
| Human | hsa-miR-362-5p | MIMAT0000705 |
Stem-loop Details
Human

Genomic Map